Learn More
MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide CN HPLC Column, 160 Å, 2.7 μm Particle Size
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Diameter (Metric) | 500 μm |
| Length (Metric) | 15 cm |
| Particle Size | 2.7 μm |
| pH | 1 to 8 |
| Pore Size | 160 Å |
| Stationary Phase | Cyano |
| Product Line | BIOshell |
| Matrix | Cyano Phase/Spherical Silica Particle Platform |
| Carbon Load | 1.5% |
| Endcapped | Yes |
| Show More |
BIOshell is a trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.