Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide CN HPLC Column, 160 Å, 2.7 μm Particle Size

Catalog No. 111007144 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
1 mm
2.1 mm
3 mm
300 μm
4.6 mm
500 μm
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-7144 10 cm 3 mm
11-100-6836 5 cm 300 μm
11-100-6614 15 cm 300 μm
11-100-6837 5 cm 500 μm
11-100-6615 15 cm 500 μm
11-100-7408 5 cm 1 mm
11-100-6873 15 cm 1 mm
11-100-8000 3 cm 2.1 mm
11-100-7559 5 cm 2.1 mm
11-100-7143 10 cm 2.1 mm
11-100-6905 15 cm 2.1 mm
11-100-7560 5 cm 3 mm
11-100-6906 15 cm 3 mm
11-100-7561 5 cm 4.6 mm
11-100-7145 10 cm 4.6 mm
11-100-6907 15 cm 4.6 mm
11-100-7374 7.5 cm 2.1 mm
11-100-8001 3 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 11-100-7144 missing translation for 'mfr' MilliporeSigma™ Supelco™ missing translation for 'supplierNo' 66972U
Add to Cart
Add to Cart

BIOshell™ 160 Å Peptide CN columns provide an alternative selectivity to the traditional C18 bonded phases.

The introduction of the cyano group enables peptides and small proteins to be separated due to a combination of hydrophobicity and dipole-dipole interactions making this column an excellent choice for biomolecules that are not resolved on traditional C18 columns. These columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected cyanopropyl ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C).
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 10 cm
Particle Size 2.7 μm
pH 1 to 8
Pore Size 160 Å
Stationary Phase Cyano
Product Line BIOshell
Matrix Cyano Phase/Spherical Silica Particle Platform
Carbon Load 1.5%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.