Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ IgG 1000 Å Diphenyl HPLC Column, 2.7 μm Particle Size

Catalog No. 111022282 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
10 cm
15 cm
25 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2282 10 cm 4.6 mm
11-102-2272 5 cm 2.1 mm
11-102-2273 10 cm 2.1 mm
11-102-2283 15 cm 4.6 mm
11-102-2274 15 cm 2.1 mm
11-102-2275 25 cm 2.1 mm
11-102-2276 3 cm 3 mm
11-102-2277 5 cm 3 mm
11-102-2278 10 cm 3 mm
11-102-2279 15 cm 3 mm
11-102-2280 3 cm 4.6 mm
11-102-2281 5 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-102-2282 Supplier MilliporeSigma™ Supelco™ Supplier No. 577429U
Add to Cart
Add to Cart

A high-speed, high-performance liquid chromatography column based on a new wide-pore (1000 Å) Fused-Core™ particle design.

  • Particle provides a thin, porous shell of high-purity silica surrounding a solid silica core
  • Particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm
  • Densely bonded, extensively endcapped diphenylmethyl bonded phase of BIOshell™ IgG 1000 Å Diphenyl provides a stable, reversed-phase packing
  • Can be used for separating high molecular weight compounds such as proteins
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 10 cm
Particle Size 2.7 μm
pH 2 to 9
Pore Size 1000 Å
Surface Area 22 m2/g
USP Type L11
Stationary Phase Diphenyl (diphenylmethyl)
Product Line BIOshell
Matrix Diphenyl (diphenylmethyl) Phase/Fused-Core™ Particle Platform
Carbon Load 1%
Endcapped Yes
Phase Reversed Phase
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.