Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek BioStack Automated Microplate Stackers, IVD PROMO

Catalog No. p-200009168 Shop All Agilent Technologies Inc. Products
Click to view available options
Description:
BioStack 3 Microplate Stacker
BioStack 4 Microplate Stacker
Plate Type:
ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, max. plate height 14.6 mm
ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates; 24- and 48-well plates (model dependent) Maximum plate height: 23.2 mm
Holds:
10 and 30 plate stacks are removable and interchangeable (50-plate stacks may be used with non-lidded plates only); 96-/384-well plates: Up to 30 plates (with lids); 1536-well plates: Up to 75 plates
10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
This item is not returnable. View return policy

Catalog No. BTBSK3WRSIP

Supplier: Agilent BioTek BSTCK3WRSI2766

This item is not returnable. View return policy

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. For In Vitro Diagnostic Use.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity

BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.

TRUSTED_SUSTAINABILITY

Specifications

Hertz 50/60 Hz
Voltage 100/240 V
Operating Temperature Range (Metric) 18°C to 40°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.7 in.
Depth (Metric) 52.6 cm
Holds 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
Humidity 10 to 85%
Includes One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
Plate Type ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, max. plate height 14.6 mm
Software LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
Weight (English) 25 lb.
Weight (Metric) 11.3 kg
Width (English) 7.4 in.
Width (Metric) 18.8 cm
Description BioStack 3 Microplate Stacker
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.