Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Filter Cubes for Imaging Multi-Mode Reader
GSA_VAGREENER_CHOICE

Catalog No. BT1225106
Click to view available options
Label or Dye:
CY5
CY7
DAPI
GFP
GFP/CFP
RFP
Texas Red
YFP
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Imaging Multi-Mode Readers.

For use with Imaging Multi-Mode Readers.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Filter Cube
For Use With (Equipment) Imaging Multi-Mode Reader
Label or Dye CY7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.