Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agilent BioTek Filter Cubes for Imaging Multi-Mode Reader



Click to view available options
Label or Dye:
CY5
CY7
DAPI
GFP
GFP/CFP
RFP
Texas Red
YFP
Description
For use with Imaging Multi-Mode Readers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Type | Filter Cube |
For Use With (Equipment) | Imaging Multi-Mode Reader |
Color | Green/Cyan |
Label or Dye | GFP/CFP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction