Learn More
Agilent BioTek Gas Controller for CO2 Control
Catalog No. bt1210007 Shop All Agilent Technologies Inc. Products
Click to view available options
For Use With (Equipment):
Cytation3 Cell Imaging multi-mode reader and synergy H1 hybrid microplate reader
This item is not returnable. View return policy
Catalog No. BT1210007
Supplier: Agilent BioTek 1210007
This item is not returnable. View return policy
For use with Cytation3 Cell Imaging multi-mode reader and synergy H1 hybrid microplate reader.
  • Compact design
  • Infrared CO2 sensor
  • Easy to read gas level indicators
  • Intuitive control panel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Controller
For Use With (Equipment) Cytation3 Cell Imaging multi-mode reader and synergy H1 hybrid microplate reader
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.