Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
GREENER_CHOICE

Catalog No. p-3543273
Click to view available options
Diameter (Metric):
0.019 cm
0.02 cm
0.035 cm
Type:
Large Volume Cassette
Large Volume Replacement Tubing Set
Low Volume 1536 Well Cassette
Low Volume Cassette
Low Volume Replacement Tubing Set
Medium Volume Cassette
Medium Volume Replacement Tubing Set
Material:
Molded Polypropylene
Stainless Steel
Capacity (Metric):
1 to 50 μL
10 to 3000 μL
5 to 2500 μL
This item is not returnable. View return policy
This item is not returnable. View return policy

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.

  • Volume range 1 μL to 10 mL
  • Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
  • Dispense 1536 wells in 18 s.
  • Cassettes can be autoclaved without recalibration
  • Dispense up to eight reagents at one time
  • Customizable dispense pattern allows skipped columns or rows
  • Built-in priming trough
  • Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
  • Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput

TRUSTED_SUSTAINABILITY

Specifications

Increments 1 μL
Material Molded Polypropylene
Capacity (Metric) 1 to 50 μL
Diameter (English) 0.01 in.
Diameter (Metric) 0.02 cm
For Use With (Application) For liquid handling
Type Low Volume Cassette
For Use With (Equipment) EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.