Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin-Anti-His-Tag Antibody

Biotin-Anti-His-Tag Antibody

Supplier:  LIFE TECHNOLOGIES PV6090

Encompass_Preferred

 View more versions of this product

Catalog No. PV6090

  • $15,492.65 / Each of 1

    Save 7%

    Reg : $16,570.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
Request a Quote

Description

Description

Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kinase Assay
Biotin
150kDa
RUO
Primary
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C.
Dry Ice
0.5 mg⁄ml
Biotin-Anti-His-Tag Antibody, 1mg, 0.5mg/mL Concentration, Biotin Conjugate, HEPES buffered saline (137mM NaCl, 2.7mM KCl, 10mM HEPES pH 7.5) Storage Buffer, Used for Pharma and Biopharma, Target and Lead Identification and Validation
1 mg
Kinase Biology
1 Each
Frozen
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.