Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Antibody (1D4-C5), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB287397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287397 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287397 Supplier Novus Biologicals Supplier No. NBP323510DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Biotin Monoclonal antibody specifically detects Biotin in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Biotin
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 1D4-C5
Conjugate DyLight 594
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias Vitamin B4
Host Species Mouse
Immunogen Biotinylated Limulus hemocyanin
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.