Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Antibody, Alkaline Phosphatase, Novus Biologicals™
SDP

Catalog No. NB1206652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206652 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206652 Supplier Novus Biologicals Supplier No. NB1206652
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Biotin Polyclonal specifically detects Biotin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Biotin
Applications Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:500-1:2500, ELISA 1:3000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Knockout Validated
Formulation 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.1% Sodium Azide
Gene Alias Vitamin B4
Host Species Goat
Immunogen Biotin conjugated to Keyhole Limpet Hemocyanin (b-KLH)
Purification Method Immunogen affinity purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling, Epitope Tags, metabolism
Primary or Secondary Primary
Test Specificity Biotin
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.