Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Antibody (BK-1/39), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP250120V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-50120V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-50120V Supplier Novus Biologicals Supplier No. NBP250120V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Biotin Monoclonal specifically detects Biotin in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Biotin
Applications Immunohistochemistry
Classification Monoclonal
Clone BK-1/39
Conjugate DyLight 405
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Vitamin B4
Host Species Mouse
Immunogen Biotin conjugated to keyhole limpet haemocyanin
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Signaling, Epitope Tags, metabolism
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.