Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BTN/8672R), Novus Biologicals™

Rabbit Monoclonal Antibody
$260.82 - $490.41
Specifications
| Antigen | Biotin |
|---|---|
| Clone | BTN/8672R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, In-situ Hybridization |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) |
Description
Biotin Monoclonal antibody specifically detects Biotin in All samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In-situ HybridizationSpecifications
| Biotin | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| Unconjugated | |
| Rabbit | |
| All species | |
| Vitamin B4 | |
| IgG κ | |
| Protein A purified |
| BTN/8672R | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, In-situ Hybridization | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| Biotinylated sheep immunoglobulin | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title