Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Antibody (BTN/8672R), Novus Biologicals™
SDP

Catalog No. NB235099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB235099 100 μg
NB233986 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB235099 Supplier Novus Biologicals Supplier No. NBP320583100UG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Biotin Monoclonal antibody specifically detects Biotin in All samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In-situ Hybridization

Specifications

Antigen Biotin
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Classification Monoclonal
Clone BTN/8672R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, In-situ Hybridization
Formulation 10mM PBS with 0.05% BSA
Gene Alias Vitamin B4
Host Species Rabbit
Immunogen Biotinylated sheep immunoglobulin
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.