Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Antibody, Rhodamine - F(ab')2, Novus Biologicals™
SDP

Catalog No. NBP19738305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19738305 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19738305 Supplier Novus Biologicals Supplier No. NBP1973830.5MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Biotin Polyclonal specifically detects Biotin in Non-species specific samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Biotin
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Rhodamine
Dilution Immunocytochemistry/Immunofluorescence 1:500-1:2500
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, 10Mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Vitamin B4
Host Species Goat
Immunogen Biotin conjugated to Keyhole Limpet Hemocyanin (b-KLH).
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Primary
Test Specificity Biotin
Reconstitution Reconstitute with 1.0mL deionized water.
Target Species Non-species specific
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.