Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin (Vitamin B7 or Vitamin H) Antibody (BTN/2032R) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB261393100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261393100 100 μg
NB261393200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB261393100 Supplier Novus Biologicals Supplier No. NBP261393100UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Biotin (Vitamin B7 or Vitamin H) Monoclonal specifically detects Biotin (Vitamin B7 or Vitamin H) in All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, In-situ Hybridization, CyTOF-ready.

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Classification Monoclonal
Clone BTN/2032R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In-situ Hybridization
Formulation 10mM PBS - BSA free with No Preservative
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Molecular Weight of Antigen 2.4 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Primary or Secondary Primary
Test Specificity It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.