Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (BTN/2032R), DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Biotin (Vitamin B7 or Vitamin H) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BTN/2032R |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H |
| Host Species | Rabbit |
| Immunogen | Biotinylated sheep immunoglobulin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.