Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375), Alexa Fluor™ 647, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Biotin (Vitamin B7 or Vitamin H) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM375 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H |
| Host Species | Mouse |
| Immunogen | Biotinylated sheep immunoglobulin |
| Molecular Weight of Antigen | 244 kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.