Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin (Vitamin B7 or Vitamin H) Mouse anti-All species, Clone: BTN399, Novus Biologicals™
SDP

Catalog No. NB177759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177759 20 μg
NB177758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177759 Supplier Novus Biologicals Supplier No. NBP31390120UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in All species samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Classification Monoclonal
Clone BTN399
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In Situ Hybridization (ISH)
Formulation 10 mM PBS with 0.05% BSA
Gene Alias 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.