Learn More
Eisco™ Biotite Gneiss Specimen
SCI_ED
Catalog No. S24780 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24780 1 Ea. 2 to 3 cm
S26586 10/Pk. 2 to 3 cm
S24781 12/Pk. 2 to 3 cm
S26635 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24780 Supplier Eisco™ Supplier No. ESNG0055
Add to Cart
Add to Cart
Biotite Gneiss is high-grade metamorphic rock formed from other rocks undergoing alteration from intense heat and pressure.
  • For use in learning about differential stresses and foliation in metamorphic rocks
  • Correlates well to Next Generation Science Standards Evidence
  • To achieve its characteristic banding, gneiss must be exposed to immense heat and pressure, and forms deep below earth
  • For use in demonstrating metamorphic properties and can be used in lessons about rock cycle
Specifications
Product Type Metamorphic Rock
For Use With (Application) Learning about differential stresses and foliation in metamorphic rocks
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Biotite Gneiss