Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BirA Antibody (5B11c3-3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP59939646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP59939646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP59939646 Supplier Novus Biologicals Supplier No. NBP259939JF646

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

BirA Monoclonal specifically detects BirA in E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BirA
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5B11c3-3
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase
Gene Symbols birA
Host Species Mouse
Immunogen GST fused to E.coli BirA R118G
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Epitope Tags
Primary or Secondary Primary
Gene ID (Entrez) 948469
Test Specificity The antibody recognises the C-terminal half of E.coli BirA; the antibody recognised the BirA variant miniTurbo; not tested for TurboID or other BirA derivate.
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.