Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BirA Antibody (6C4c7), FITC, Novus Biologicals™
SDP

Catalog No. NBP259938F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259938F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259938F Supplier Novus Biologicals Supplier No. NBP259938F

Mouse Monoclonal Antibody

BirA Monoclonal specifically detects BirA in E. coli, Yeast samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BirA
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6C4c7
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase
Gene Symbols birA
Host Species Mouse
Immunogen GST fused to E.coli BirA R118G
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Epitope Tags
Primary or Secondary Primary
Gene ID (Entrez) 948469
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.