Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC8 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176669 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176669 Supplier Novus Biologicals Supplier No. NBP176669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BIRC8 Polyclonal specifically detects BIRC8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BIRC8
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_203127
Gene Alias baculoviral IAP repeat containing 8, baculoviral IAP repeat-containing 8, baculoviral IAP repeat-containing protein 8, hILP2, IAP-like protein 2, ILP-2ILP2, Inhibitor of apoptosis-like protein 2, Testis-specific inhibitor of apoptosis
Gene Symbols BIRC8
Host Species Rabbit
Immunogen Antibody was raised with against a synthetic peptide corresponding to amino acids near the amino terminus of human ILP-2. The immunogen is located within the first 50 amino acids of ILP-2. Amino Acid Squence: TGYEARLITFGT
Molecular Weight of Antigen 33 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 112401
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.