Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Birch pollen Bet V1 Antibody (102H10.05), Novus Biologicals™
SDP

Catalog No. DDX0050P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0050P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0050P100 Supplier Novus Biologicals Supplier No. DDX0050P100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Block/Neutralize, Western Blot.

Specifications

Antigen Birch pollen Bet V1
Applications Blocking Assay, Western Blot
Classification Monoclonal
Clone 102H10.05
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Block/Neutralize
Gene Alias Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Host Species Mouse
Immunogen Birch Pollen
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Pollen.
Target Species Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.