Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Birch pollen Bet V1 Antibody (BV16), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB015895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015895 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015895 Supplier Novus Biologicals Supplier No. NBP281093AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Birch pollen Bet V1
Applications Western Blot, ELISA
Classification Monoclonal
Clone BV16
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Host Species Rabbit
Immunogen Birch pollen Bet V1-A, a major allergen of the European white birch.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody binds to Birch pollen Bet V1.
Target Species Plant
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.