Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(2-butoxyethyl) Ether, 98.5%, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Specifications
Specifications
Acidity | 100 ppm |
CAS Max % | 1 |
Packaging | Drum |
Quantity | 200 L |
Percent Purity | 0.985 |
Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction