Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(2-hydroxyethyl) disulfide, tech. 90%
Click to view available options
Quantity:
50 g
250 g
Description
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.
Solubility
Soluble in water, ethanol, acetone, and ether.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.
Solubility
Soluble in water, ethanol, acetone, and ether.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | ∼25°C |
| Density | 1.261 |
| Boiling Point | 158°C to 163°C (3.5 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Unpleasant |
| Refractive Index | 1.57 |
| Linear Formula | HOCH2CH2S-SCH2CH2OH |
| Quantity | 50 g |
| UN Number | UN2810 |
| Beilstein | 1735851 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H318 |
| Hazard Statement | GHS H Statement H301-H318 Toxic if swallowed. Causes serious eye damage. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 217-576-6 |
| RTECSNumber | KK7525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction