Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol, 99%, extra pure

Catalog No. p-3735633
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 111-46-6 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
PubChem CID 8117
ChEBI CHEBI:46807
SMILES OCCOCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -10.0°C
Density 1.1100g/mL
Boiling Point 245.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4480
Linear Formula (HOCH2CH2)2O
Quantity 1 L
Beilstein 01, 468
Specific Gravity 1.11
Merck Index 15, 3131
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, ether,, ethylene glycol, insoluble in benzene, toluene, carbon, tetrachloride and aliphatic hydrocarbons
IUPAC Name 2-(2-hydroxyethoxy)ethan-1-ol
Viscosity 38 mPa.s (20°C)
Formula Weight 106.12
Percent Purity 99%
Grade Extra Pure
Physical Form Oily Liquid
Chemical Name or Material Diethylene glycol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 203-872-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.