Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, AcroSeal™

Catalog No. AC428981000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 202289-38-1 | C6H14F3NO2S | 221.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [Bis(2-methoxyethyl)amino]sulfur trifluoride
CAS 202289-38-1, 108-88-3
Color Orange-Yellow
Density 1.2000g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C6H14F3NO2S
Linear Formula (CH3OCHCH2)2NSF3
Quantity 100 mL
MDL Number MFCD01321415
Specific Gravity 1.2
Merck Index 15, 2899
Synonym bis 2-methoxyethyl aminosulfur trifluoride, deoxo-fluor, 2-methoxy-n-2-methoxyethyl-n-trifluorothio-ethanamine, deoxo-fluor r, bis 2-methoxyethyl aminosulphurtrifluoride, bast, bis 2-methoxyethyl amino sulfur trifluoride, bis 2-methoxyethyl aminosulphur trifluoride
InChI Key APOYTRAZFJURPB-UHFFFAOYSA-N
SMILES COCCN(CCOC)S(F)(F)F
IUPAC Name 2-methoxy-N-(2-methoxyethyl)-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
Molecular Weight (g/mol) 221.24
PubChem CID 2734690
Formula Weight 221.24
Percent Purity ≥95% (NMR, neat substance)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.