Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-bromophenyl) Sulfide 98.0+%, TCI America™
SDP

Catalog No. p-7123536 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44351G 1 g
B44355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44351G Supplier TCI America Supplier No. B44351G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3393-78-0
Molecular Formula C12H8Br2S
Molecular Weight (g/mol) 344.064
InChI Key CLPVCAXOQZIJGW-UHFFFAOYSA-N
PubChem CID 76929
IUPAC Name 1-bromo-4-(4-bromophenyl)sulfanylbenzene
SMILES C1=CC(=CC=C1SC2=CC=C(C=C2)Br)Br

Specifications

Melting Point 115°C
Color White
Boiling Point 210°C
Quantity 1 g
Formula Weight 344.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl) Sulfide
RTECSNumber WP9900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.