Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-chlorophenyl) sulfone, 99%

Catalog No. p-7024381
Click to view available options
Quantity:
100 g
500 g

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol

Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 147°C to 150°C
Boiling Point 249°C to 251°C (10 mmHg)
Flash Point 233°C (451°F)
Quantity 100 g
Beilstein 2052955
Solubility Information Insoluble in water.
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
Formula Weight 287.17
Percent Purity 99%
Chemical Name or Material Bis(4-chlorophenyl) sulfone
Hazard Category H319-H500
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-247-9
RTECSNumber WR3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.