Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-hydroxyphenyl) sulfone, 99%

Catalog No. p-7023608
Click to view available options
Quantity:
250 g
1000 g

CAS: 80-09-1 | C12H10O4S | 250.27 g/mol

It is a reactant used in epoxy reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant used in epoxy reactions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00002350
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
PubChem CID 6626
ChEBI CHEBI:34372
IUPAC Name 4-(4-hydroxyphenyl)sulfonylphenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 245°C to 250°C
Density 1.366
Quantity 250 g
Beilstein 2052954
Solubility Information Insoluble in water.
Formula Weight 250.28
Percent Purity 99%
Chemical Name or Material Bis(4-hydroxyphenyl) sulfone
Hazard Category H319-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-250-5
RTECSNumber SM8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.