Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-methoxyphenyl)disulfide, 97%

Catalog No. aab2539903
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAB2539903

Supplier: Thermo Scientific Chemicals B2539903

Only null left
Add to Cart

CAS: 5335-87-5 | C14H14O2S2 | 278.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5335-87-5
Molecular Formula C14H14O2S2
Molecular Weight (g/mol) 278.38
MDL Number MFCD00041358
InChI Key PZQGLCGLPMWYBT-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl disulfide, disulfide, bis 4-methoxyphenyl, p-methoxyphenyl disulfide, 4,4'-dimethoxy diphenyldisulfide, 4-methoxyphenyl disulfide, anisole, 4,4'-dithiodi, 1-methoxy-4-4-methoxyphenyl disulfanyl benzene, 1,2-bis 4-methoxyphenyl disulfane, bis p-methoxyphenyl disulfide
PubChem CID 79258
IUPAC Name 1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene
SMILES COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 37°C to 41°C
Boiling Point 200°C (3 mmHg)
Quantity 1 g
Beilstein 1914789
Formula Weight 278.39
Percent Purity 97%
Chemical Name or Material Bis(4-methoxyphenyl)disulfide
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.