Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
887919-35-9
C32H56Cl2N2P2Pd
708.07
MFCD09265123
DWOZNANUEDYIOF-UHFFFAOYSA-L
bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii,pd amphos cl2,pdcl2 amphos 2,a-taphos 2pdcl2,dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii,alpha-taphos 2pdcl2,bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride
11714597
4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.Cl[Pd]Cl
887919-35-9
C32H56Cl2N2P2Pd
708.07
MFCD09265123
DWOZNANUEDYIOF-UHFFFAOYSA-L
bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii,pd amphos cl2,pdcl2 amphos 2,a-taphos 2pdcl2,dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii,alpha-taphos 2pdcl2,bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride
11714597
4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.Cl[Pd]Cl
Packaging Glass bottle
Quantity 5g
Assay Percent Range 94% min. (HPLC)
Formula Weight 708.07
Percent Purity 95%
Chemical Name or Material Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning