Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(phenylsulfonyl)sulfide, 96%

Catalog No. p-4510205
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 4388-22-1 | C12H10O4S3 | 314.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4388-22-1
Molecular Formula C12H10O4S3
Molecular Weight (g/mol) 314.39
MDL Number MFCD00094733
InChI Key YQUSJUJNDKUWAM-UHFFFAOYSA-N
Synonym bis phenylsulfonyl sulfide, benzenesulfonyl sulfide, benzenesulfonic thioanhydride, benzenesulfonothioic acid, anhydrosulfide, benzenesulfonic acid, thio-, anhydrosulfide, bis phenylsulfonyl thio, benzenesulfonyl sulfanylsulfonyl benzene, phso2 2s, benzenesulfonylthio sulfonylbenzene
PubChem CID 257516
IUPAC Name benzenesulfonylsulfanylsulfonylbenzene
SMILES O=S(=O)(SS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 129.0°C to 134.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 11,III,163
Formula Weight 314.41
Percent Purity 96%
Physical Form Crystals or Powder
Chemical Name or Material Bis(phenylsulfonyl)sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.