Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Citrate, Powder, USP, 49-54%, Spectrum™ Chemical
SDP

Catalog No. 18600999 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

B1973, 813-93-4, C6H5BiO7

Spectrum™ Chemical Bismuth Citrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 813-93-4
Molecular Formula C6H5BiO7
Molecular Weight (g/mol) 398.08
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
IUPAC Name bismuth(3+) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 49% to 54%
Grade USP
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.