Learn More
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)
Description
In semiconductor research, multilayer thin-film, magnetoresistant film.Bismuth(III) selenide is used as a semiconductor and thermoelectric material. It is also used in semiconductor research. Further, it finds applications magnetoresistant film and semiconductor research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 710°C |
| Color | Gray |
| Physical Form | Pieces |
| Quantity | 10 g |
| UN Number | UN3283 |
| Merck Index | 14,1278 |
| Solubility Information | Insoluble in water; Decomposes when heated in air; Decomposes in nitric acid and aqua regiaSoluble in strong acids; Insoluble in water and organic solvents. |
| Formula Weight | 654.84 |
| Percent Purity | 99.999% |
| Density | 6.82 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
| EINECSNumber | 235-104-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.