Alfa Aesar™ Bismuth(III) selenide, Vacuum Deposition Grade, 99.999% (metals basis)

In semiconductor research, multilayer thin-film, magnetoresistant film

Manufacturer: Alfa Aesar™ 1312609

Quantity 10g

 View more versions of this product

Catalog No. AA1312609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12068-69-8
    Formula Weight 654.84
    MDL Number MFCD00014200
    UN Number UN3283
    Merck Index 14,1278
    Chemical Name or Material Bismuth(III) selenide
    Grade Vacuum Deposition Grade

    Specifications

    Formula Weight 654.84
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size 1 to 6mm
    CAS 12068-69-8
    UN Number UN3283
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    MDL Number MFCD00014200
    Solubility Information Insoluble in water; Decomposes when heated in air; Decomposes in nitric acid and aqua regia
    Health Hazard 1 Danger
    Density 6.82g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
    Physical Form Pieces
    Merck Index 14,1278
    Color Gray
    TSCA TSCA
    Quantity 10g
    Melting Point 710°C
    Chemical Name or Material Bismuth(III) selenide
    Assay 99.999%, (metals basis)
    Grade Vacuum Deposition Grade

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit