Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Lead Tin Cadmium ingot (Wood's metal)

Catalog No. AA33218A3
Click to view available options
Quantity:
100 g
500 g
2 kg

Catalog No. AA33218A3

Supplier: Thermo Scientific Chemicals 033218A3

Only null left
Add to Cart

CAS: 8049-22-7 | Cd12Sn2 | 1586.388 g/mol

Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8049-22-7
Molecular Formula Cd12Sn2
Molecular Weight (g/mol) 1586.388
MDL Number MFCD00198175
InChI Key OIBLPQFTAJDRMC-UHFFFAOYSA-N
Synonym lambda∼2∼-stannane-cadmium 1/6
PubChem CID 71434152
IUPAC Name cadmium;tin
SMILES [Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn]

Specifications

Color Gray
Melting Point 71°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt%
Quantity 2 kg
Odor Odorless
Form Ingot
UN Number UN3077
Solubility Information Insoluble in water.
Assay (Wood's metal)
Chemical Name or Material Bismuth Lead Tin Cadmium ingot (Wood's metal)
Hazard Category H302-H330-H341-H350-H360Df-H372
Hazard Statement GHS H Statement
H330-H350-H360-H372-H373-H341-H302-H201
Fatal if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May cause damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Explosive
mass explosion hazard.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.