Learn More
Bismuth Lead Tin Cadmium ingot (Wood's metal)
Description
Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Melting Point | 71°C |
| Avg. Mol. Wt. or Mol. Wt. Range | Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt% |
| Quantity | 500 g |
| Odor | Odorless |
| Form | Ingot |
| UN Number | UN3077 |
| Solubility Information | Insoluble in water. |
| Assay | (Wood's metal) |
| Chemical Name or Material | Bismuth Lead Tin Cadmium ingot (Wood's metal) |
Safety and Handling
| Hazard Category | H302-H330-H341-H350-H360Df-H372 |
| Hazard Statement | GHS H Statement H330-H350-H360-H372-H373-H341-H302-H201 Fatal if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Harmful if swallowed. Explosive mass explosion hazard. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P304+P340-P308+P313-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.