Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subnitrate (Powder/USP), Fisher Chemical

Catalog No. b344500 Shop All Fisherbrand Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B344-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. B344-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 1304-85-4
Molecular Formula Bi5H9N4O22
MDL Number MFCD00064844
Synonym Bismuth Nitrate, Basic, Bismuth Nitrate, Oxide, Bismuth Oxynitrate

Specifications

Melting Point 500°C
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 500 g
Linear Formula Bi5O(OH)9(NO3)4
Formula Weight 1461.99
Percent Purity ≥79.0% (USP)
Grade USP
Loss on Drying 3.0% max. (USP)
Packaging Poly Bottle
Chemical Name or Material Bismuth Subnitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.