Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIX 01294
GREENER_CHOICE

Catalog No. 336450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GLP and G9a inhibitor

GLP and G9a histone lysine methyltransferase inhibitor (IC50 values are 0.7 and 1.7 μM respectively) that displays no activity at other histone methyltransferases up to 37 μM. Modulates H3K9me2 levels in mammalian cells and potentiates induction of pluripotent stem cells from somatic cells in vitro. Also inhibits H3K36 methylation by oncoproteins NSD1, NSD2 and NSD3 (IC50 values are 40 - 112 μM).

Chemical Identifiers

CAS 1392399-03-9
Molecular Formula C28H41Cl3N6O2

Specifications

Quantity 50 mg
Formula Weight 600.02
Purity >98%
Molecular Weight (g/mol) 600.03
Chemical Name or Material BIX 01294
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.