Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BIX 02189

Click to view available options
Quantity:
10 mg
Description
BIX 02189 is a selective MEK5 and ERK5 inhibitor (IC50 values are 1.5 and 59 nM respectively). Inhibits sorbitol-induced ERK5 phosphorylation in HeLa cells. Selective for MEK5 and ERK5 over MEK1/2, ERK1/2 and a panel of 87 kinases. Induces apoptosis in acute myeloid leukemia tumor cells.
Specifications
Specifications
Quantity | 10 mg |
Product Type | BIX 02189 |
Purity | >98% |
Molecular Weight (g/mol) | 440.54 |
Inhibitors | BIX 02189 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction