Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BJ Long™ Pipe Cleaner Brushes

Catalog No. p-26063
Encompass
Click to view available options
For Use With (Equipment):
0.098 to 0.196 in. O.D. Tubing

Cut at any length for cleaning small pipets, stopcocks, capillary tubes

  • May be used for slightly larger sizes by bending in moderate zig-zag pattern

Specifications

Product Type Pipe Cleaner Brush
Handle Material Galvanized Wire
For Use With (Application) For cleaning small pipets, stopcocks, capillary tubes
For Use With (Equipment) 0.098 to 0.196 in. O.D. Tubing
Length (English) 53 ft.
Length (Metric) 16.2 m
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.