Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BL 1249
GREENER_CHOICESDP

Catalog No. 379710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-971-0 10 mg
37-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-971-0 Supplier Tocris Bioscience™ Supplier No. 3797/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Putative K2P2.1 (TREK-1) channel opener

Putative potassium channel activator; thought to act as a K2P2.1 (TREK-1) channel opener. Exhibits selectivity for bladder over vascular tissue in vitro and in vivo (EC50 values are 1.26 and 21.0 μM for cultured bladder and aortic tissues respectively).

Chemical Identifiers

CAS 18200-13-0
Molecular Formula C17H17N5

Specifications

Quantity 10 mg
Formula Weight 291.35
Purity >99%
Molecular Weight (g/mol) 291.358
Chemical Name or Material BL 1249
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.