Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BL21-lysate Rabbit anti-Bacteria, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | BL21-lysate |
|---|---|
| Concentration | 0.7 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6404
![]()
|
Proteintech Group Inc
105571IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Escherichia coli B serves as a research model for studying phage sensitivity, restriction-modification systems, and bacterial evolution, and also as a workhorse for protein expression in life science laboratories and in the biotech industry. Characteristics such as protease deficiency, low acetate production at a high level of glucose, and enhanced permeability (probably due to a simple cell surface) make E. coli B a desirable host for the production of genetically engineered proteins. BL21(DE3) has been very widely used to express recombit proteins. Comparison of BL21(DE3) and another B strain REL606 shows they differ in length by 72. 304 bp and have 426 single nucleotide polymorphisms, all of which have been accounted for by various laboratory manipulations. Differences between B strains and K12 include the absence of flagellar component genes, the DNA cytosine methylase dcm, and ompT in BL21(DE3). B strains may have an additional type II secretion system not found in K12. Other differences include differential distribution of insertions sequences and other changes resulting from horizontal gene transfer. BL21(DE3) also carries a DE3 recombit phage harboring the T7 RNA polymerase gene that can direct high-level expression of cloned genes under the control of the T7 promoter.Specifications
| BL21-lysate | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| BL21 lysate | |
| IgG | |
| Protein A | |
| Antibody |
| 0.7 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BL21-lysate Fusion Protein Ag0049 | |
| Primary | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title