Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLAME/SLAMF8 Antibody (210), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB076140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076140 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076140 Supplier Novus Biologicals Supplier No. NBP290656G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BLAME/SLAMF8 Monoclonal specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for ELISA.

Specifications

Antigen BLAME/SLAMF8
Applications ELISA
Classification Monoclonal
Clone 210
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BLAME/SLAMF8 (NP_083360.2; Met1-Asp231).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56833
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.