Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bleomycin Sulfate, PROMO, Thermo Scientific Chemicals

Catalog No. J60727MAPM
Click to view available options
Quantity:
10 mg

Bleomycin sulfate is a glycopeptide compound with antibiotic activity isolated from Streptomyces verticillus. It also exhibits anticancer activity.

General Description

  • Bleomycin sulfate was initially isolated from Streptomyces verticillus. It is a glycopeptide compound with antibiotic activity
  • It has the ability to induce oxidative cleavage of the DNA backbone, leading to apoptosis
  • Bleomycin sulfate can form a complex with iron that reduces oxygen to superoxide and hydroxyl radicals. These reactive oxygen species can cause single- and double-stranded breaks in DNA as well as promote carbohydrate oxidation, lipid peroxidation, and alterations in prostaglandin synthesis and degradation

Application

  • It can be used to inhibit DNA metabolism and slow or stop cell growth. This property explains its anticancer activity
  • Bleomycin sulfate can also be used to degrade RNA as it is more selective to RNA than DNA
  • Local injections of bleomycin sulfate in mouse models can induce sclerotic skin mimicking scleroderma
  • In laboratory research, bleomycin is used to induce pulmonary fibrosis in mice
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-93-4
Molecular Formula C55H85N17O25S4
Molecular Weight (g/mol) 1512.62 g/mol
MDL Number MFCD00070310
InChI Key WUIABRMSWOKTOF-UHFFFAOYSA-N
PubChem CID 131664136
IUPAC Name [2-[2-[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[5-[[1-[2-[4-[4-(3-dimethylsulfoniopropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hyd
SMILES CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)OC4C(C(C(C(O4)CO)O)OC(=N)[O-])O)C(=O)NC(C)C(C(C)C(=O)NC(C(C)O)C(=O)NCCC5=NC(=CS5)C6=NC(=CS6)C(=O)NCCC[S+](C)C)O.OS(=O)(=O)O

Specifications

Type Bleomycin Sulfate
Physical Form Powder
Color White
Quantity 10 mg
Merck Index 14,1318
Solubility Information Soluble in water or DMSO
Chemical Name or Material Bleomycin sulfate
EINECSNumber 232-925-2
RTECSNumber EC5991990
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.