Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLIMP1/PRDM1 Antibody (3H2-E8), HRP, Novus Biologicals™
SDP

Catalog No. NB600235H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600235H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600235H Supplier Novus Biologicals Supplier No. NB600235H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human, Mouse, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen BLIMP1/PRDM1
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3H2-E8
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. O75626
Gene Alias Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1
Gene Symbols PRDM1
Host Species Mouse
Immunogen A fragment of mouse Blimp-1 corresponding to residues 199-409. [UniProt# Q60636]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 639
Target Species Human, Mouse, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.