Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLIMP1/PRDM1 Antibody (646702), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB348282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348282 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348282 Supplier Novus Biologicals Supplier No. FAB36081T
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLIMP1/PRDM1 Monoclonal antibody specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry

Specifications

Antigen BLIMP1/PRDM1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 646702
Conjugate Alexa Fluor 594
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human BLIMP1/PRDM1, Lys667-Cys789, Accession # O75626
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 639
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.